Merck
  • Toward the Standardization of Mitochondrial Proteomics: The Italian Mitochondrial Human Proteome Project Initiative.
Toward the Standardization of Mitochondrial Proteomics: The Italian Mitochondrial Human Proteome Project Initiative.
Journal of proteome research (2017-08-23)
Tiziana Alberio, Luisa Pieroni, Maurizio Ronci, Cristina Banfi, Italia Bongarzone, Patrizia Bottoni, Maura Brioschi, Marianna Caterino, Clizia Chinello, Antonella Cormio, Flora Cozzolino, Vincenzo Cunsolo, Simona Fontana, Barbara Garavaglia, Laura Giusti, Viviana Greco, Antonio Lucacchini, Elisa Maffioli, Fulvio Magni, Francesca Monteleone, Maria Monti, Valentina Monti, Clara Musicco, Giuseppe Petrosillo, Vito Porcelli, Rosaria Saletti, Roberto Scatena, Alessio Soggiu, Gabriella Tedeschi, Mara Zilocchi, Paola Roncada, Andrea Urbani, Mauro Fasano
ABSTRACT
The Mitochondrial Human Proteome Project aims at understanding the function of the mitochondrial proteome and its crosstalk with the proteome of other organelles. Being able to choose a suitable and validated enrichment protocol of functional mitochondria, based on the specific needs of the downstream proteomics analysis, would greatly help the researchers in the field. Mitochondrial fractions from ten model cell lines were prepared using three enrichment protocols and analyzed on seven different LC-MS/MS platforms. All data were processed using neXtProt as reference database. The data are available for the Human Proteome Project purposes through the ProteomeXchange Consortium with the identifier PXD007053. The processed data sets were analyzed using a suite of R routines to perform a statistical analysis and to retrieve subcellular and submitochondrial localizations. Although the overall number of identified total and mitochondrial proteins was not significantly dependent on the enrichment protocol, specific line to line differences were observed. Moreover, the protein lists were mapped to a network representing the functional mitochondrial proteome, encompassing mitochondrial proteins and their first interactors. More than 80% of the identified proteins resulted in nodes of this network but with a different ability in coisolating mitochondria-associated structures for each enrichment protocol/cell line pair.
Sigma-Aldrich
Mitochondria Isolation Kit, sufficient for 50 applications (2-5 x 107 cells), isolation of enriched mitochondrial fraction from cells
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution