Merck
  • RNF8 up-regulates AR/ARV7 action to contribute to advanced prostate cancer progression.
RNF8 up-regulates AR/ARV7 action to contribute to advanced prostate cancer progression.
Cell death & disease (2022-04-17)
Tingting Zhou, Shengli Wang, Xiaoyu Song, Wensu Liu, Fang Dong, Yunlong Huo, Renlong Zou, Chunyu Wang, Siyi Zhang, Wei Liu, Ge Sun, Lin Lin, Kai Zeng, Xiang Dong, Qiqiang Guo, Fei Yi, Zhuo Wang, Xiaoman Li, Bo Jiang, Liu Cao, Yue Zhao
ABSTRACT
Androgen receptor (AR) signaling drives prostate cancer (PC) progression. Androgen deprivation therapy (ADT) is temporally effective, whereas drug resistance inevitably develops. Abnormal expression of AR/ARV7 (the most common AR splicing variant) is critical for endocrine resistance, while the detailed mechanism is still elusive. In this study, bioinformatics and immunohistochemical analyses demonstrate that RNF8 is high expressed in PC and castration-resistant PC (CRPC) samples and the expression of RNF8 is positively correlated with the Gleason score. The high expression of RNF8 in PCs predicts a poor prognosis. These results provide a potential function of RNF8 in PC progression. Furthermore, the mRNA expression of RNF8 is positively correlated with that of AR in PC. Mechanistically, we find that RNF8 upregulates c-Myc-induced AR transcription via altering histone modifications at the c-Myc binding site within the AR gene. RNF8 also acts as a co-activator of AR, promoting the recruitment of AR/ARV7 to the KLK3 (PSA) promoter, where RNF8 modulates histone modifications. These functions of RNF8 are dependent on its E3 ligase activity. RNF8 knockdown further reduces AR transactivation and PSA expression in CRPC cells with enzalutamide treatment. RNF8 depletion restrains cell proliferation and alleviates enzalutamide resistance in CRPC cells. Our findings indicate that RNF8 may be a potential therapeutic target for endocrine resistance in PC.
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse