Merck
  • Validation of a small molecule inhibitor of PDE6D-RAS interaction with favorable anti-leukemic effects.
Validation of a small molecule inhibitor of PDE6D-RAS interaction with favorable anti-leukemic effects.
Blood cancer journal (2022-04-16)
Sara Canovas Nunes, Serena De Vita, Andrew Anighoro, François Autelitano, Edward Beaumont, Pamela Klingbeil, Meaghan McGuinness, Beatrice Duvert, Chad Harris, Lu Yang, Sheela Pangeni Pokharel, Chun-Wei Chen, Monika Ermann, David A Williams, Haiming Xu
ABSTRACT
RAS mutations prevalent in high-risk leukemia have been linked to relapse and chemotherapy resistance. Efforts to directly target RAS proteins have been largely unsuccessful. However, since RAS-mediated transformation is dependent on signaling through the RAS-related C3 botulinum toxin substrate (RAC) small GTPase, we hypothesized that targeting RAC may be an effective therapeutic approach in RAS mutated tumors. Here we describe multiple small molecules capable of inhibiting RAC activation in acute lymphoblastic leukemia cell lines. One of these, DW0254, also demonstrates promising anti-leukemic activity in RAS-mutated cells. Using chemical proteomics and biophysical methods, we identified the hydrophobic pocket of phosphodiester 6 subunit delta (PDE6D), a known RAS chaperone, as a target for this compound. Inhibition of RAS localization to the plasma membrane upon DW0254 treatment is associated with RAC inhibition through a phosphatidylinositol-3-kinase/AKT-dependent mechanism. Our findings provide new insights into the importance of PDE6D-mediated transport for RAS-dependent RAC activation and leukemic cell survival.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Ras Antibody, (K-, H-, N-), clone 9A11.2, clone 9A11.2, Upstate®, from mouse
Sigma-Aldrich
Ras Activation Assay Kit