Merck
  • Succinyl-CoA Ligase Deficiency in Pro-inflammatory and Tissue-Invasive T Cells.
Succinyl-CoA Ligase Deficiency in Pro-inflammatory and Tissue-Invasive T Cells.
Cell metabolism (2020-12-03)
Bowen Wu, Jingtao Qiu, Tuantuan V Zhao, Yanan Wang, Toshihisa Maeda, Isabel N Goronzy, Mitsuhiro Akiyama, Shozo Ohtsuki, Ke Jin, Lu Tian, Jörg J Goronzy, Cornelia M Weyand
ABSTRACT
Autoimmune T cells in rheumatoid arthritis (RA) have a defect in mitochondrial oxygen consumption and ATP production. Here, we identified suppression of the GDP-forming β subunit of succinate-CoA ligase (SUCLG2) as an underlying abnormality. SUCLG2-deficient T cells reverted the tricarboxylic acid (TCA) cycle from the oxidative to the reductive direction, accumulated α-ketoglutarate, citrate, and acetyl-CoA (AcCoA), and differentiated into pro-inflammatory effector cells. In AcCoAhi RA T cells, tubulin acetylation stabilized the microtubule cytoskeleton and positioned mitochondria in a perinuclear location, resulting in cellular polarization, uropod formation, T cell migration, and tissue invasion. In the tissue, SUCLG2-deficient T cells functioned as cytokine-producing effector cells and were hyperinflammatory, a defect correctable by replenishing the enzyme. Preventing T cell tubulin acetylation by tubulin acetyltransferase knockdown was sufficient to inhibit synovitis. These data link mitochondrial failure and AcCoA oversupply to autoimmune tissue inflammation.
Sigma-Aldrich
CTP Inhibitor, ≥98% (HPLC)