Merck
  • Endothelial-Mesenchymal Transition in Heart Failure With a Preserved Ejection Fraction: Insights Into the Cardiorenal Syndrome.
Endothelial-Mesenchymal Transition in Heart Failure With a Preserved Ejection Fraction: Insights Into the Cardiorenal Syndrome.
Circulation. Heart failure (2021-08-20)
María Valero-Muñoz, Albin Oh, Elizabeth Faudoa, Rosa Bretón-Romero, Fatima El Adili, Andreea Bujor, Flora Sam
ABSTRACT
The management of clinical heart failure with a preserved ejection fraction (HFpEF) is often complicated by concurrent renal dysfunction, known as the cardiorenal syndrome. This, combined with the notable lack of evidence-based therapies for HFpEF, highlights the importance of examining mechanisms and targetable pathways in HFpEF with the cardiorenal syndrome. HFpEF was induced in mice by uninephrectomy, infusion of d-aldosterone (HFpEF; N=10) or saline (Sham; N=8), and given 1% NaCl drinking water for 4 weeks. Renal fibrosis and endothelial-mesenchymal transition (endo-MT) were evident once HFpEF developed. Human aortic endothelial cells were treated for 4 days with 10% serum obtained from patients with chronically stable HFpEF with the cardiorenal syndrome (N=12) and compared with serum-treated human aortic endothelial cells from control subjects (no cardiac/renal disease; N=12) to recapitulate the in vivo findings. Kidneys from HFpEF mice demonstrated hypertrophy, interstitial fibrosis (1.9-fold increase; P<0.05) with increased expression of endo-MT transcripts, including pdgfrβ (platelet-derived growth factor receptor β), snail, fibronectin, fsp1 (fibroblast-specific protein 1), and vimentin by 1.7- (P=0.004), 1.7- (P=0.05), 1.8- (P=0.005), 2.6- (P=0.001), and 2.0-fold (P=0.001) versus Sham. Immunostaining demonstrated co-localization of CD31 and ACTA2 (actin α2) in kidney sections suggesting evidence of endo-MT. Similar to the findings in HFpEF mice, comparable endo-MT markers were also significantly elevated in human aortic endothelial cells treated with serum from patients with HFpEF compared with human aortic endothelial cells treated with serum from control subjects. These translational findings demonstrate a plausible role for endo-MT in HFpEF with cardiorenal syndrome and may have therapeutic implications in drug development for patients with HFpEF and concomitant renal dysfunction.
Sigma-Aldrich
Anti-PDGFR Beta Antibody, from rabbit, purified by affinity chromatography