- Viral Kinetics of an Acute Hepatitis B Virus Subgenotype F1b Infection in a Mexican Subject.
Viral Kinetics of an Acute Hepatitis B Virus Subgenotype F1b Infection in a Mexican Subject.
Clinical liver disease (2022-03-22)
Arturo Panduro, Sonia Roman, Nora A Fierro, João R Rebello-Pinho
PMID35308473
ABSTRACT
Content available: Author Interview and Audio Recording.