Merck
  • Nanohydroxyapatite Hydrogel Can Promote the Proliferation and Migration of Chondrocytes and Better Repair Talar Articular Cartilage.
Nanohydroxyapatite Hydrogel Can Promote the Proliferation and Migration of Chondrocytes and Better Repair Talar Articular Cartilage.
Computational and mathematical methods in medicine (2022-06-07)
Yuxuan Zhang, Yi Cui, Jian Tian, Xueming Chen, Tonglong Xu, Jiajia Liu, Yajun Xu
ABSTRACT
As an important load-bearing part of the body, joints are prone to articular cartilage degradation during exercise, resulting in joint pain, swelling, and deformity, which has an adverse impact on patients' life quality and social medical security. Therefore, this study aims to test an effective biopolymer scaffold in promoting the growth of chondrocytes in talus. Hydrogel (Gel)-nanohydroxyapatite (nHA) was invented as a new type of biopolymer scaffold for osteoarthritis treatment in this research. To detect the effects of Gel-nHA on guidance, cartilage matrix secretion, mineralization, proliferation, and migration of chondrocyte, we cultured chondrocytes to study the biological properties of nHA. It was found that Gel could guide chondrocytes to permeate and migrate, so it could be used as acellular matrix scaffolds for chondrocyte regeneration. In addition, nHA could stimulate chondrocytes to secrete cartilage matrix, such as type II collagen and mucopolysaccharide (GAGs). At the same time, nHA help to induce chondrocyte mineralization and stimulate the secretion of type X collagen, so as to better maintain the integrity of bone cartilage interface. In Gel-nHA, chondrocyte viability could be better maintained, and the proliferation and migration of chondrocytes could be better promoted, so as to better repair the articular cartilage of talus. Therefore, the Gel-nHA scaffold is expected to become an effective method for repairing talus cartilage in the future.