Merck
  • A look into retinal organoids: methods, analytical techniques, and applications.
A look into retinal organoids: methods, analytical techniques, and applications.
Cellular and molecular life sciences : CMLS (2021-08-23)
Tess A V Afanasyeva, Julio C Corral-Serrano, Alejandro Garanto, Ronald Roepman, Michael E Cheetham, Rob W J Collin
ABSTRACT
Inherited retinal diseases (IRDs) cause progressive loss of light-sensitive photoreceptors in the eye and can lead to blindness. Gene-based therapies for IRDs have shown remarkable progress in the past decade, but the vast majority of forms remain untreatable. In the era of personalised medicine, induced pluripotent stem cells (iPSCs) emerge as a valuable system for cell replacement and to model IRD because they retain the specific patient genome and can differentiate into any adult cell type. Three-dimensional (3D) iPSCs-derived retina-like tissue called retinal organoid contains all major retina-specific cell types: amacrine, bipolar, horizontal, retinal ganglion cells, Müller glia, as well as rod and cone photoreceptors. Here, we describe the main applications of retinal organoids and provide a comprehensive overview of the state-of-art analysis methods that apply to this model system. Finally, we will discuss the outlook for improvements that would bring the cellular model a step closer to become an established system in research and treatment development of IRDs.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Aquaporin 1 Antibody, 1.0 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Prox 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Rhodopsin Antibody, clone 4D2, clone 4D2, from mouse
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid
Sigma-Aldrich
Anti-PAX6 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Calbindin D-28K (Ab-1) Rabbit pAb, lyophilized, Calbiochem®
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Opsin Antibody, Red/Green, Chemicon®, from rabbit
Sigma-Aldrich
Anti-G Protein Goα Antibody, clone 2A, clone 2A, Chemicon®, from mouse
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 7E3-1B8, clone 7E3-1B8, Chemicon®, from mouse
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Dopamine β Hydroxylase Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Rhodopsin Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Prospero homeobox protein 1/PROX1 Antibody, from rabbit, purified by affinity chromatography