Merck
  • Deficiency in endocannabinoid synthase DAGLB contributes to early onset Parkinsonism and murine nigral dopaminergic neuron dysfunction.
Deficiency in endocannabinoid synthase DAGLB contributes to early onset Parkinsonism and murine nigral dopaminergic neuron dysfunction.
Nature communications (2022-06-18)
Zhenhua Liu, Nannan Yang, Jie Dong, Wotu Tian, Lisa Chang, Jinghong Ma, Jifeng Guo, Jieqiong Tan, Ao Dong, Kaikai He, Jingheng Zhou, Resat Cinar, Junbing Wu, Armando G Salinas, Lixin Sun, Mantosh Kumar, Breanna T Sullivan, Braden B Oldham, Vanessa Pitz, Mary B Makarious, Jinhui Ding, Justin Kung, Chengsong Xie, Sarah L Hawes, Lupeng Wang, Tao Wang, Piu Chan, Zhuohua Zhang, Weidong Le, Shengdi Chen, David M Lovinger, Cornelis Blauwendraat, Andrew B Singleton, Guohong Cui, Yulong Li, Huaibin Cai, Beisha Tang
ABSTRACT
Endocannabinoid (eCB), 2-arachidonoyl-glycerol (2-AG), the most abundant eCB in the brain, regulates diverse neural functions. Here we linked multiple homozygous loss-of-function mutations in 2-AG synthase diacylglycerol lipase β (DAGLB) to an early onset autosomal recessive Parkinsonism. DAGLB is the main 2-AG synthase in human and mouse substantia nigra (SN) dopaminergic neurons (DANs). In mice, the SN 2-AG levels were markedly correlated with motor performance during locomotor skill acquisition. Genetic knockdown of Daglb in nigral DANs substantially reduced SN 2-AG levels and impaired locomotor skill learning, particularly the across-session learning. Conversely, pharmacological inhibition of 2-AG degradation increased nigral 2-AG levels, DAN activity and dopamine release and rescued the locomotor skill learning deficits. Together, we demonstrate that DAGLB-deficiency contributes to the pathogenesis of Parkinsonism, reveal the importance of DAGLB-mediated 2-AG biosynthesis in nigral DANs in regulating neuronal activity and dopamine release, and suggest potential benefits of 2-AG augmentation in alleviating Parkinsonism.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)