Merck
  • Calcineurin regulates the stability and activity of estrogen receptor α.
Calcineurin regulates the stability and activity of estrogen receptor α.
Proceedings of the National Academy of Sciences of the United States of America (2021-10-30)
Takahiro Masaki, Makoto Habara, Yuki Sato, Takahiro Goshima, Keisuke Maeda, Shunsuke Hanaki, Midori Shimada
ABSTRACT
Estrogen receptor α (ER-α) mediates estrogen-dependent cancer progression and is expressed in most breast cancer cells. However, the molecular mechanisms underlying the regulation of the cellular abundance and activity of ER-α remain unclear. We here show that the protein phosphatase calcineurin regulates both ER-α stability and activity in human breast cancer cells. Calcineurin depletion or inhibition down-regulated the abundance of ER-α by promoting its polyubiquitination and degradation. Calcineurin inhibition also promoted the binding of ER-α to the E3 ubiquitin ligase E6AP, and calcineurin mediated the dephosphorylation of ER-α at Ser294 in vitro. Moreover, the ER-α (S294A) mutant was more stable and activated the expression of ER-α target genes to a greater extent compared with the wild-type protein, whereas the extents of its interaction with E6AP and polyubiquitination were attenuated. These results suggest that the phosphorylation of ER-α at Ser294 promotes its binding to E6AP and consequent degradation. Calcineurin was also found to be required for the phosphorylation of ER-α at Ser118 by mechanistic target of rapamycin complex 1 and the consequent activation of ER-α in response to β-estradiol treatment. Our study thus indicates that calcineurin controls both the stability and activity of ER-α by regulating its phosphorylation at Ser294 and Ser118 Finally, the expression of the calcineurin A-α gene (PPP3CA) was associated with poor prognosis in ER-α-positive breast cancer patients treated with tamoxifen or other endocrine therapeutic agents. Calcineurin is thus a promising target for the development of therapies for ER-α-positive breast cancer.
Sigma-Aldrich
Calmodulin, His•Tag® Human, Recombinant, Calmodulin, His•Tag Human, Recombinant, is a full-length, recombinant human calmodulin fused to a His•Tag sequence at the N-terminus. Contains four functional Ca2+-binding sites.