Merck
  • Targeting CRABP-II overcomes pancreatic cancer drug resistance by reversing lipid raft cholesterol accumulation and AKT survival signaling.
Targeting CRABP-II overcomes pancreatic cancer drug resistance by reversing lipid raft cholesterol accumulation and AKT survival signaling.
Journal of experimental & clinical cancer research : CR (2022-03-10)
Shuiliang Yu, Lei Wang, Danian Che, Mei Zhang, Ming Li, Mikihiko Naito, Wei Xin, Lan Zhou
ABSTRACT
Resistance to standard therapy is a major reason for the poor prognosis of pancreatic ductal adenocarcinoma (PDAC). Developing novel therapy to overcome PDAC drug-resistance is urgently needed. CRABP-II was highly expressed in all PDAC but not expressed in normal pancreatic tissues and chronic pancreatitis. CRABP-II was shown to promote PDAC migration and metastasis while its potential role in promoting PDAC drug-resistance was not known. A paired cohort of human primary and relapsing PDAC tissues was assessed for CRABP-II expression by immunohistochemistry. CRISPR/cas9 gene editing was used to establish CRABP-II knockout cell lines and MTT assays were performed to assess gemcitabine sensitivity in vitro. Cleaved caspase-3/PARP blots and Annexin V staining were conducted to detect cell apoptosis. Gene expression microarray, Q-PCR, western blots, Co-IP and RNA-IP were used to study the molecular function of CRABP-II. Sucrose gradient ultracentrifugation was applied to isolate lipid rafts and LC-MS-MS was used to assess cholesterol content. Both subcutaneous CDX models and orthotopic PDX models were established to examine the efficacy of SNIPER-11 and the synergistic effect between SNIPER-11 and gemcitabine in vivo. A higher expression of CRABP-II was found in relapsing PDAC tissue and was associated with poor prognosis. Gemcitabine-resistant cell lines exhibited increased level of CRABP-II, while CRABP-II knockout resensitized PDAC cells to gemcitabine. Mechanistically, aberrant expression of CRABP-II increased the stability of SREBP-1c mRNA through cooperation with HuR and upregulated the downstream genes of SREBP-1c to favor cholesterol uptake and accumulation in lipid rafts. Increased lipid raft cholesterol accumulation facilitated ATK survival signaling and PDAC drug resistance. The small compound SNIPER-11 treatment effectively induced CRABP-II protein degradation, induced apoptosis, and suppressed tumor growth. Combination of SNIPER-11 and gemcitabine significantly reduced the lipid raft cholesterol content in CDX/PDX and profoundly inhibited tumor progression. These findings identified CRABP-II as a novel regulator of cholesterol metabolism and suggested that CRABP-II is a selective target for overcoming PDAC drug resistance.
Sigma-Aldrich
Anti-CRABP2 Antibody, ascites fluid, Chemicon®