- Evaluation of long-chain fatty acid respiration in neonatal mouse cardiomyocytes using SeaHorse instrument.
Evaluation of long-chain fatty acid respiration in neonatal mouse cardiomyocytes using SeaHorse instrument.
STAR protocols (2022-05-24)
Aude Angelini, Xinchun Pi, Liang Xie
PMID35600933
ABSTRACT
Metabolic switches play a critical role in the pathophysiology of cardiac diseases, including heart failure. Here, we describe an assay for long-chain fatty acid oxidation in neonatal mouse cardiomyocytes by using a SeaHorse Flux Analyzer (Agilent). This protocol is a simplified but robust adaptation of the standard protocol that enables metabolic measurements in cells isolated from transgenic mouse models, which can be timesaving and informative. Cell isolation and culture represent a critical point that may require bench optimization. For complete details on the use and execution of this protocol, please refer to Angelini et al. (2021).