Merck
  • Transcriptional Regulation of Amino Acid Transport in Glioblastoma Multiforme.
Transcriptional Regulation of Amino Acid Transport in Glioblastoma Multiforme.
Cancers (2021-12-25)
Robyn A Umans, Joelle Martin, Megan E Harrigan, Dipan C Patel, Lata Chaunsali, Aarash Roshandel, Kavya Iyer, Michael D Powell, Ken Oestreich, Harald Sontheimer
ABSTRACT
Glioblastoma multiforme (GBM) is a deadly brain tumor with a large unmet therapeutic need. Here, we tested the hypothesis that wild-type p53 is a negative transcriptional regulator of SLC7A11, the gene encoding the System xc- (SXC) catalytic subunit, xCT, in GBM. We demonstrate that xCT expression is inversely correlated with p53 expression in patient tissue. Using representative patient derived (PDX) tumor xenolines with wild-type, null, and mutant p53 we show that p53 expression negatively correlates with xCT expression. Using chromatin immunoprecipitation studies, we present a molecular interaction whereby p53 binds to the SLC7A11 promoter, suppressing gene expression in PDX GBM cells. Accordingly, genetic knockdown of p53 increases SLC7A11 transcript levels; conversely, over-expressing p53 in p53-null GBM cells downregulates xCT expression and glutamate release. Proof of principal studies in mice with flank gliomas demonstrate that daily treatment with the mutant p53 reactivator, PRIMA-1Met, results in reduced tumor growth associated with reduced xCT expression. These findings suggest that p53 is a molecular switch for GBM glutamate biology, with potential therapeutic utility.
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution