Merck
  • RNA splicing is a key mediator of tumour cell plasticity and a therapeutic vulnerability in colorectal cancer.
RNA splicing is a key mediator of tumour cell plasticity and a therapeutic vulnerability in colorectal cancer.
Nature communications (2022-05-20)
Adam E Hall, Sebastian Öther-Gee Pohl, Patrizia Cammareri, Stuart Aitken, Nicholas T Younger, Michela Raponi, Caroline V Billard, Alfonso Bolado Carrancio, Aslihan Bastem, Paz Freile, Fiona Haward, Ian R Adams, Javier F Caceres, Paula Preyzner, Alex von Kriegsheim, Malcolm G Dunlop, Farhat V Din, Kevin B Myant
ABSTRACT
Tumour cell plasticity is a major barrier to the efficacy of targeted cancer therapies but the mechanisms that mediate it are poorly understood. Here, we identify dysregulated RNA splicing as a key driver of tumour cell dedifferentiation in colorectal cancer (CRC). We find that Apc-deficient CRC cells have dysregulated RNA splicing machinery and exhibit global rewiring of RNA splicing. We show that the splicing factor SRSF1 controls the plasticity of tumour cells by controlling Kras splicing and is required for CRC invasion in a mouse model of carcinogenesis. SRSF1 expression maintains stemness in human CRC organoids and correlates with cancer stem cell marker expression in human tumours. Crucially, partial genetic downregulation of Srsf1 does not detrimentally affect normal tissue homeostasis, demonstrating that tumour cell plasticity can be differentially targeted. Thus, our findings link dysregulation of the RNA splicing machinery and control of tumour cell plasticity.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid