Merck
  • CEP97 phosphorylation by Dyrk1a is critical for centriole separation during multiciliogenesis.
CEP97 phosphorylation by Dyrk1a is critical for centriole separation during multiciliogenesis.
The Journal of cell biology (2021-11-18)
Moonsup Lee, Kunio Nagashima, Jaeho Yoon, Jian Sun, Ziqiu Wang, Christina Carpenter, Hyun-Kyung Lee, Yoo-Seok Hwang, Christopher J Westlake, Ira O Daar
ABSTRACT
Proper cilia formation in multiciliated cells (MCCs) is necessary for appropriate embryonic development and homeostasis. Multicilia share many structural characteristics with monocilia and primary cilia, but there are still significant gaps in our understanding of the regulation of multiciliogenesis. Using the Xenopus embryo, we show that CEP97, which is known as a negative regulator of primary cilia formation, interacts with dual specificity tyrosine phosphorylation regulated kinase 1A (Dyrk1a) to modulate multiciliogenesis. We show that Dyrk1a phosphorylates CEP97, which in turn promotes the recruitment of Polo-like kinase 1 (Plk1), which is a critical regulator of MCC maturation that functions to enhance centriole disengagement in cooperation with the enzyme Separase. Knockdown of either CEP97 or Dyrk1a disrupts cilia formation and centriole disengagement in MCCs, but this defect is rescued by overexpression of Separase. Thus, our study reveals that Dyrk1a and CEP97 coordinate with Plk1 to promote Separase function to properly form multicilia in vertebrate MCCs.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Thrombin Protease, Cytiva 27-0846-01
Sigma-Aldrich
Rosetta(DE3) Competent Cells - Novagen, Rosetta host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Sigma-Aldrich
Anti-Centrin Antibody, clone 20H5, clone 20H5, from mouse