Merck
  • The Potential Tumor-Suppressor DHRS7 Inversely Correlates with EGFR Expression in Prostate Cancer Cells and Tumor Samples.
The Potential Tumor-Suppressor DHRS7 Inversely Correlates with EGFR Expression in Prostate Cancer Cells and Tumor Samples.
Cancers (2022-07-10)
Simon Stücheli, Selene Araya, Caner Ercan, Seraina O Moser, John Gallon, Paul Jenö, Salvatore Piscuoglio, Luigi Terracciano, Alex Odermatt
ABSTRACT
Prostate cancer (PCa), one of the most common malignancies in men, typically responds to initial treatment, but resistance to therapy often leads to metastases and death. The dehydrogenase/reductase 7 (DHRS7, SDR34C1) is an "orphan" enzyme without known physiological function. DHRS7 was previously found to be decreased in higher-stage PCa, and siRNA-mediated knockdown increased the aggressiveness of LNCaP cells. To further explore the role of DHRS7 in PCa, we analyzed the proteome of LNCaP cells following DHRS7 knockdown to assess potentially altered pathways. Although DHRS7 is able to inactivate 5α-dihydrotestosterone, DHRS7 knockdown did not affect androgen receptor (AR) target gene expression, and its effect on PCa cells seems to be androgen-independent. Importantly, proteome analyses revealed increased expression of epidermal growth factor receptor (EGFR), which was confirmed by RT-qPCR and Western blotting. Comparison of AR-positive LNCaP with AR-negative PC-3 and DU145 PCa cell lines revealed a negative correlation between DHRS7 and EGFR expression. Conversely, EGFR knockdown enhanced DHRS7 expression in these cells. Importantly, analysis of patient samples revealed a negative correlation between DHRS7 and EGFR expression, both at the mRNA and protein levels, and DHRS7 expression correlated positively with patient survival rates. These results suggest a protective role for DHRS7 in PCa.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture