Merck
  • Time-resolved phosphoproteome and proteome analysis reveals kinase signaling on master transcription factors during myogenesis.
Time-resolved phosphoproteome and proteome analysis reveals kinase signaling on master transcription factors during myogenesis.
iScience (2022-06-21)
Di Xiao, Marissa Caldow, Hani Jieun Kim, Ronnie Blazev, Rene Koopman, Deborah Manandi, Benjamin L Parker, Pengyi Yang
ABSTRACT
Myogenesis is governed by signaling networks that are tightly regulated in a time-dependent manner. Although different protein kinases have been identified, knowledge of the global signaling networks and their downstream substrates during myogenesis remains incomplete. Here, we map the myogenic differentiation of C2C12 cells using phosphoproteomics and proteomics. From these data, we infer global kinase activity and predict the substrates that are involved in myogenesis. We found that multiple mitogen-activated protein kinases (MAPKs) mark the initial wave of signaling cascades. Further phosphoproteomic and proteomic profiling with MAPK1/3 and MAPK8/9 specific inhibitions unveil their shared and distinctive roles in myogenesis. Lastly, we identified and validated the transcription factor nuclear factor 1 X-type (NFIX) as a novel MAPK1/3 substrate and demonstrated the functional impact of NFIX phosphorylation on myogenesis. Altogether, these data characterize the dynamics, interactions, and downstream control of kinase signaling networks during myogenesis on a global scale.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Supelco
SYPRO® Ruby Protein Gel Stain