Merck
  • A highly selective near-infrared fluorescent probe for carboxylesterase 2 and its bioimaging applications in living cells and animals.
A highly selective near-infrared fluorescent probe for carboxylesterase 2 and its bioimaging applications in living cells and animals.
Biosensors & bioelectronics (2016-04-30)
Qiang Jin, Lei Feng, Dan-Dan Wang, Jing-Jing Wu, Jie Hou, Zi-Ru Dai, Shi-Guo Sun, Jia-Yue Wang, Guang-Bo Ge, Jing-Nan Cui, Ling Yang
ABSTRACT
A near-infrared fluorescent probe (DDAB) for highly selective and sensitive detection of carboxylesterase 2 (CE2) has been designed, synthesized, and systematically studied both in vitro and in vivo. Upon addition of CE2, the ester bond of DDAB could be rapidly cleaved and then release a near-infrared (NIR) fluorophore DDAO, which brings a remarkable yellow-to-blue color change and strong NIR fluorescence emission in physiological solutions. The newly developed probe exhibits excellent properties including good specificity, ultrahigh sensitivity and high imaging resolution. Moreover, DDAB has been applied to measure the real activities of CE2 in complex biological samples, as well as to screen CE2 inhibitors by using tissue preparations as the enzymes sources. The probe has also been successfully used to detect endogenous CE2 in living cells and in vivo for the first time, and the results demonstrate that such detection is highly reliable. All these prominent features of DDAB make it holds great promise for further investigation on CE2-associated biological process and for exploring the physiological functions of CE2 in living systems.
Sigma-Aldrich
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes