Merck
  • High expression of monocarboxylate transporter 4 (MCT 4), but not MCT 1, predicts poor prognosis in patients with non-small cell lung cancer.
High expression of monocarboxylate transporter 4 (MCT 4), but not MCT 1, predicts poor prognosis in patients with non-small cell lung cancer.
Translational cancer research (2022-02-05)
Ying-Hui Tong, Xiao-Ping Hu, Xue-Ping Xiang, Luo Fang
ABSTRACT
The monocarboxylate transporter (MCT) family especially MCT1 and MCT4 have been recognized to play an important role in lactate transport, a key glycolytic product. The expression of MCT1 and MCT4 expression was previously found to be related to poor outcome in various cancer types. In this study, we investigated the expression status of MCT1 and MCT4 and their relationship with prognosis in non-small cell lung cancer (NSCLC). Expression of MCT4 and MCT1 in NSCLC tumor and adjacent lung tissues were detected by immunohistochemistry. Kaplan-Meier plots were used to evaluate two proteins' prognostic role, and the log-rank test obtained the P value. For multivariate analysis, the Cox proportional-hazards regression method was performed. High MCT4 and MCT1 expression was observed in cancer cells, with a rate of 45% for MCT4 versus 15% for MCT1 among all NSCLC patients. High expression of MCT4, and not MCT1, was associated with worse overall survival (OS) [hazard ratio (HR) =1.96 (1.06-3.75), P=0.032] and progression-free survival (PFS) [HR =1.72 (1.05-2.93), P=0.032] in NSCLC patients. In our multivariate analysis, advanced cancer stage and high MCT4 level were identified as independent predictive indicators for both PFS [HR(MCT4) =1.888 (1.114-3.199), P=0.018 and OS [HR (MCT4) =2.421 (1.271-4.610), P=0.007]. Subgroup and interaction analyses were also performed in different clinical characteristic groups and no significant differences were observed. High MCT4 expression is a predictive marker for worse outcome in NSCLC patients.