Merck
  • The Effectiveness of the Nimali Variety of Sri Lankan Punica granatum L. Fruit Extracts on Rhabdomyosarcoma (RD) Cells Concerning the Apoptotic Signaling Pathway.
The Effectiveness of the Nimali Variety of Sri Lankan Punica granatum L. Fruit Extracts on Rhabdomyosarcoma (RD) Cells Concerning the Apoptotic Signaling Pathway.
Asian Pacific journal of cancer prevention : APJCP (2022-03-01)
Udeshika Yapa Bandara, Chamindri Witharana, Preethi Soysa
ABSTRACT
Pomegranate ,a polyphenol-rich fruit, has been considered as one of the ancient fruits with anticancer effect. Cell cycle arrest is considered as an ordinary factor in human cancer, and apoptosis is the frequent drug target. This study aimed to evaluate the effectiveness of the Nimali variety of Sri Lankan Punica granatum L. fruit extracts on rhabdomyosarcoma (RD) cells concerning the apoptotic signaling pathway. Antiproliferative activity of aqueous extracts of pomegranate peel, pericarp, was assessed using multiple extraction methods (sonication, microwaving, sonication followed by microwaving, keeping in a waterbath, and boiling at 100ºC). Total protein content, nitric oxide production, LDH, and caspase-8 and caspase-3 activities were analyzed in peel extracts prepared by sonicated or microwave methods. RT-qPCR was performed with intact RNA to explore the apoptotic pathway and gene expression. Peel extracts expressed minimum cell viability in a dose-dependent manner, induced cell death on RD cells. However, sonicated peel extract (SPL) indicated the lowest IC50 of 14.8±2.2 μg/mL comparative to healthy VERO cells (>1,000 μg/mL). A decrease of nitrite content in the supernatant was visualized in the graph plotted against concentration. Furthermore, SPL upregulated caspase-8 and caspase-3 signaling pathways and expression of p21 and p53 genes. The findings highlighted the promising therapeutic potential of SPL to inhibit RD growth and progression and to modulate the caspase-8 and caspase-3, p53, and p21 dependent pathway.
Sigma-Aldrich
Lactate Dehydrogenase Activity Assay Kit, sufficient for 500 colorimetric tests
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Fetal Bovine Serum, Heat Inactivated, non-USA origin, sterile-filtered, suitable for cell culture