Merck
  • High extracellular Ca2+ stimulates Ca2+-activated Cl- currents in frog parathyroid cells through the mediation of arachidonic acid cascade.
High extracellular Ca2+ stimulates Ca2+-activated Cl- currents in frog parathyroid cells through the mediation of arachidonic acid cascade.
PloS one (2011-05-12)
Yukio Okada, Kotapola G Imendra, Toshihiro Miyazaki, Hitoshi Hotokezaka, Rie Fujiyama, Kazuo Toda
ABSTRACT
Elevation of extracellular Ca(2+) concentration induces intracellular Ca(2+) signaling in parathyroid cells. The response is due to stimulation of the phospholipase C/Ca(2+) pathways, but the direct mechanism responsible for the rise of intracellular Ca(2+) concentration has remained elusive. Here, we describe the electrophysiological property associated with intracellular Ca(2+) signaling in frog parathyroid cells and show that Ca(2+)-activated Cl(-) channels are activated by intracellular Ca(2+) increase through an inositol 1,4,5-trisphophate (IP(3))-independent pathway. High extracellular Ca(2+) induced an outwardly-rectifying conductance in a dose-dependent manner (EC(50) ∼6 mM). The conductance was composed of an instantaneous time-independent component and a slowly activating time-dependent component and displayed a deactivating inward tail current. Extracellular Ca(2+)-induced and Ca(2+) dialysis-induced currents reversed at the equilibrium potential of Cl(-) and were inhibited by niflumic acid (a specific blocker of Ca(2+)-activated Cl(-) channel). Gramicidin-perforated whole-cell recording displayed the shift of the reversal potential in extracellular Ca(2+)-induced current, suggesting the change of intracellular Cl(-) concentration in a few minutes. Extracellular Ca(2+)-induced currents displayed a moderate dependency on guanosine triphosphate (GTP). All blockers for phospholipase C, diacylglycerol (DAG) lipase, monoacylglycerol (MAG) lipase and lipoxygenase inhibited extracellular Ca(2+)-induced current. IP(3) dialysis failed to induce conductance increase, but 2-arachidonoylglycerol (2-AG), arachidonic acid and 12S-hydroperoxy-5Z,8Z,10E,14Z-eicosatetraenoic acid (12(S)-HPETE) dialysis increased the conductance identical to extracellular Ca(2+)-induced conductance. These results indicate that high extracellular Ca(2+) raises intracellular Ca(2+) concentration through the DAG lipase/lipoxygenase pathway, resulting in the activation of Cl(-) conductance.
Sigma-Aldrich
Niflumic acid
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phorbol 12,13-dibutyrate, ≥98% (TLC), powder
Sigma-Aldrich
Orlistat, ≥98%, solid
Supelco
10 mm/10 mm Union, PTFE, pk of 3
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
1-Oleoyl-2-acetyl-sn-glycerol, ≥97% (TLC), oil
Sigma-Aldrich
5,8,11,14-Eicosatetraynoic acid, ≥97%
Sigma-Aldrich
Papain from papaya latex, crude powder, 1.5-10 units/mg solid
Sigma-Aldrich
Papain from papaya latex, lyophilized powder, aseptically filled
Sigma-Aldrich
Ruthenium Red, Technical grade
Sigma-Aldrich
Papain from papaya latex, buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein
Sigma-Aldrich
U-73122 hydrate, powder
Sigma-Aldrich
Papain from Carica papaya, solution, light brown, ≥10 U/mg protein (~25 mg/ml)