Merck
Complement consumption in children with Plasmodium falciparum malaria.
Malaria journal (2009-01-13)
Nancy K Nyakoe, Ronald P Taylor, Joseph N Makumi, John N Waitumbi
ABSTRACT
Complement (C) can be activated during malaria, C components consumed and inflammatory mediators produced. This has potential to impair host innate defence. In a case-control study, C activation was assessed by measuring serum haemolytic activity (CH50), functional activity of each pathway and levels of C3a, C4a and C5a in children presenting at Kisumu District Hospital, western Kenya, with severe malarial anaemia (SMA) or uncomplicated malaria (UM). CH50 median titers for lysis of sensitized sheep erythrocytes in SMA (8.6 U/mL) were below normal (34-70 U/mL) and were one-fourth the level in UM (34.6 U/mL (P < 0.001). Plasma C3a median levels were 10 times higher than in normals forSMA (3,200 ng/ml) and for UM (3,500 ng/ml), indicating substantial C activation in both groups. Similar trends were obtained for C4a and C5a. The activities of all three C pathways were greatly reduced in SMA compared to UM (9.9% vs 83.4% for CP, 0.09% vs 30.7% for MBL and 36.8% vs 87.7% for AP respectively, P < 0.001). These results indicate that, while C activation occurs in both SMA and UM, C consumption is excessive in SMA. It is speculated that in SMA, consumption of C exceeds its regeneration.
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, Special High EEO