Merck
Collection of Monoclonal Antibodies Targeting SARS-CoV-2 Proteins.
Viruses (2022-02-27)
Marina Pribanić Matešić, Paola Kučan Brlić, Tihana Lenac Roviš, Željka Mačak Šafranko, Abigael Eva Chaouat, Karmela Miklić, Suzana Malić, Nina Ivanković, Maren Schubert, Federico Bertoglio, Alemka Markotić, Ofer Mandelboim, Stipan Jonjić, Ilija Brizić
ABSTRACT
In early 2020, the COVID-19 pandemic sparked a global crisis that continues to pose a serious threat to human health and the economy. Further advancement in research is necessary and requires the availability of quality molecular tools, including monoclonal antibodies. Here, we present the development and characterization of a collection of over 40 new monoclonal antibodies directed against different SARS-CoV-2 proteins. Recombinant SARS-CoV-2 proteins were expressed, purified, and used as immunogens. Upon development of specific hybridomas, the obtained monoclonal antibody (mAb) clones were tested for binding to recombinant proteins and infected cells. We generated mAbs against structural proteins, the Spike and Nucleocapsid protein, several non-structural proteins (nsp1, nsp7, nsp8, nsp9, nsp10, nsp16) and accessory factors (ORF3a, ORF9b) applicable in flow cytometry, immunofluorescence, or Western blot. Our collection of mAbs provides a set of novel, highly specific tools that will allow a comprehensive analysis of the viral proteome, which will allow further understanding of SARS-CoV-2 pathogenesis and the design of therapeutic strategies.
Sigma-Aldrich
o-Phenylenediamine dihydrochloride, tablet, 30 mg substrate per tablet