Merck
  • Evaluating the susceptibility of AGO2-loaded microRNAs to degradation by nucleases in vitro.
Evaluating the susceptibility of AGO2-loaded microRNAs to degradation by nucleases in vitro.
Methods (San Diego, Calif.) (2018-05-20)
Reyad A Elbarbary, Lynne E Maquat
ABSTRACT
MicroRNAs (miRNAs) comprise a class of small non-coding RNAs that regulate the stability and/or translatability of most protein-coding transcripts. Steady-state levels of mature miRNAs can be controlled through mechanisms that influence their biogenesis and/or decay rates. Pathways that mediate mature miRNA decay are less well understood than those that mediate miRNA biogenesis. We recently described Tudor-staphylococcal/micrococcal-like nuclease (TSN)-mediated miRNA decay (TumiD) as a cellular pathway that promotes the sequence-specific endonucleolytic decay of miRNAs that harbor a CA and/or UA dinucleotide. Here, we describe an in vitro assay for evaluating the susceptibility of AGO2-loaded miRNAs to degradation by different classes of nucleases. This in vitro approach can be used to complement in vivo studies that aim to identify novel miRNA decay factors.
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt solution, HPLC purified, aqueous solution for RNA polymerase transcription
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial