Merck
  • NF-κB directly regulates β-arrestin-1 expression and forms a negative feedback circuit in TNF-α-induced cell death.
NF-κB directly regulates β-arrestin-1 expression and forms a negative feedback circuit in TNF-α-induced cell death.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-02-28)
Juan Li, Ao Guo, Qinying Wang, Yuanyuan Li, Jian Zhao, Jing Lu, Gang Pei
ABSTRACT
β-Arrestins (β-arrestin-1 and -2) are multifunctional proteins that play important roles in the regulation of inflammation and cell survival that need to be tightly controlled; however, the mechanism that underlies their gene expression is largely unclear. Here, we demonstrate that β-arrestin-1 is a transcriptional target of NF-κB. mRNA and protein levels of β-arrestin-1 were up-regulated by NF-κB inducers. Inhibition of NF-κB prevented the up-regulation of β-arrestin-1 mRNA, whereas activation of NF-κB led to increased β-arrestin-1 expression. β-Arrestin-1 promoter activity was consistently enhanced upon NF-κB activation as a result of the presence of a highly conserved κB site. β-Arrestin-1, in turn, suppressed the transcriptional activity of NF-κB by interfering with the interaction between p65 and p50. β-Arrestin-1-deficient mice displayed reduced TNF-α-induced cell death and increased expression of antiapoptotic genes. Reintroduction of β-arrestin-1, but not its mutant, which is unable to interfere with the p65-p50 interaction, into β-arrestin-deficient mouse embryonic fibroblasts partially restored sensitivity to TNF-α-induced cell death. These findings reveal NF-κB and β-arrestin-1 to be key components of a negative feedback circuit that is necessary to regulate cell death.-Li, J., Guo, A., Wang, Q., Li, Y., Zhao, J., Lu, J., Pei, G. NF-κB directly regulates β-arrestin-1 expression and forms a negative feedback circuit in TNF-α-induced cell death.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
JumpStart Taq ReadyMix for Quantitative PCR, For probe-based real-time PCR
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection