Merck
  • Fluorometric Estimation of Glutathione in Cultured Microglial Cell Lysate.
Fluorometric Estimation of Glutathione in Cultured Microglial Cell Lysate.
Bio-protocol (2017-06-05)
Vikas Singh, Ruchi Gera, Mahaveer Prasad Purohit, Satyakam Patnaik, Debabrata Ghosh
ABSTRACT
Glutathione is one of the major antioxidant defense components present in cells. It is predominantly present as reduced glutathione (GSH) and converted into oxidized glutathione (GSSG) while reducing the free radicals like hydroxyl ions (OH-). For the measurement of GSH and GSSG, o-phthalaldehyde (OPT) has been used as a fluorescent reagent. O-phthalaldehyde has an ability to react specifically with GSH at pH 8 and GSSG at pH 12 respectively. N-ethylmaleimide (NEM) has been used to prevent auto-oxidation of GSH during measurement of GSSG in the present protocol. The original protocol by Hissin and Hilf was developed for glutathione estimation in Rat liver tissue. The present protocol has been standardized following Hissin and Hilf (1976) for the estimation of glutathione in cultured microglial cell lysate but it can also be used for other mammalian cell lysate. In our lab same protocol has been used for the estimation of glutathione in the whole cell lysate of murine neuroblastoma cell, N2a.
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
SAFC
Dulbecco′s Modified Eagle′s Medium/Ham′s Nutrient Mixture F12, powder, with 3151 mg/L dextrose, with 2.5 mM L-glutamine, with 55 mg/L sodium pyruvate, without sodium bicarbonate
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%