Merck
  • Rapid multiresidue determination of pesticides in livestock muscle and liver tissue via modified QuEChERS sample preparation and LC-MS/MS.
Rapid multiresidue determination of pesticides in livestock muscle and liver tissue via modified QuEChERS sample preparation and LC-MS/MS.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2017-04-14)
K Hamamoto, K Iwatsuki, R Akama, R Koike
ABSTRACT
Many multiresidue methods for the determination of pesticides in vegetables and fruits have been reported to date. However, few such methods have been employed to investigate pesticide residues in animal tissue. In this study, an LC-MS/MS multiresidue method coupled with modified QuEChERS extraction was developed and validated for the investigation of eight pesticide residues: prallethrin (PR), resmethrin (RMT), imidacloprid (IMC), diflubenzuron (DFB), cyromazine (CYR), etofenprox (EFP), dinotefuran (DNT) and phthalthrin (PTLT). This method involves initial extraction in a water/acetone system, the addition of salts and a subsequent extraction/partitioning step and, finally, a clean-up step utilising dispersive solid-phase extraction (SPE). The mean recoveries of seven of the pesticides (the exception being CYR) ranged between 74.7% and 113.5%, and the CVs of the livestock tissue - bovine, swine, and chicken muscle and liver tissue spiked at 10 ng g-1 (50 ng g-1 for RMT and DNT) and 100 ng g-1 - were < 13.8%. The recoveries of CYR in all muscle and liver spiked samples ranged from 56.9% to 78.3%, while those of RMT in swine liver were > 120%. Therefore, this method was considered as being unsuitable for the investigation of these samples. The limits of quantitation (LOQs) of seven of the investigated pesticides (the exception being swine liver) in the tissue samples ranged from 0.9 to 15.2 ng g-1. We therefore concluded that this LC-MS/MS multiresidue method is a valid and suitable for the investigation of seven pesticides in animal tissue, but it is unsuitable for the analysis of CYR in all animal tissues and RMT in swine liver tissue.
Product Description
Supelco
Prallethrin, mixture of stereoisomers, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland