Merck
  • Development of a Selective CDK7 Covalent Inhibitor Reveals Predominant Cell-Cycle Phenotype.
Development of a Selective CDK7 Covalent Inhibitor Reveals Predominant Cell-Cycle Phenotype.
Cell chemical biology (2019-03-25)
Calla M Olson, Yanke Liang, Alan Leggett, Woojun D Park, Lianbo Li, Caitlin E Mills, Selma Z Elsarrag, Scott B Ficarro, Tinghu Zhang, Robert Düster, Matthias Geyer, Taebo Sim, Jarrod A Marto, Peter K Sorger, Ken D Westover, Charles Y Lin, Nicholas Kwiatkowski, Nathanael S Gray
ABSTRACT
Cyclin-dependent kinase 7 (CDK7) regulates both cell cycle and transcription, but its precise role remains elusive. We previously described THZ1, a CDK7 inhibitor, which dramatically inhibits superenhancer-associated gene expression. However, potent CDK12/13 off-target activity obscured CDK7s contribution to this phenotype. Here, we describe the discovery of a highly selective covalent CDK7 inhibitor. YKL-5-124 causes arrest at the G1/S transition and inhibition of E2F-driven gene expression; these effects are rescued by a CDK7 mutant unable to covalently engage YKL-5-124, demonstrating on-target specificity. Unlike THZ1, treatment with YKL-5-124 resulted in no change to RNA polymerase II C-terminal domain phosphorylation; however, inhibition could be reconstituted by combining YKL-5-124 and THZ531, a selective CDK12/13 inhibitor, revealing potential redundancies in CDK control of gene transcription. These findings highlight the importance of CDK7/12/13 polypharmacology for anti-cancer activity of THZ1 and posit that selective inhibition of CDK7 may be useful for treatment of cancers marked by E2F misregulation.
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-7) Antibody, clone 4E12, culture supernatant, clone 4E12, from rat
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho-CTD Ser-5) Antibody, clone 3E8, clone 3E8, from rat
Sigma-Aldrich
Anti-RNA polymerase II subunit B1 (phospho CTD Ser-2) Antibody, clone 3E10, clone 3E10, from rat
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial