Merck
  • Chain-like assembly of gold nanoparticles on artificial DNA templates via 'click chemistry'.
Chain-like assembly of gold nanoparticles on artificial DNA templates via 'click chemistry'.
Chemical communications (Cambridge, England) (2007-12-20)
Monika Fischler, Alla Sologubenko, Joachim Mayer, Guido Clever, Glenn Burley, Johannes Gierlich, Thomas Carell, Ulrich Simon
ABSTRACT
We present a new type of azide-functionalized gold nanoparticle and their coupling to an alkyne-modified DNA duplex using the copper(I)-catalyzed Huisgen cycloaddition ('click chemistry'), resulting in a chain-like assembly of nanoparticles on the DNA template.
Sigma-Aldrich
Gold nanoparticles, 20 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 70 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 5 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 90 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 15 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 80 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 40 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 30 nm diameter, azide functionalized, 1ml, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 50 nm diameter, azide functionalized, 1 ML, OD 50, in water
Sigma-Aldrich
Gold nanoparticles, 60 nm diameter, azide functionalized, 1ml, OD 50, in water