Merck
  • Neurturin is a PGC-1α1-controlled myokine that promotes motor neuron recruitment and neuromuscular junction formation.
Neurturin is a PGC-1α1-controlled myokine that promotes motor neuron recruitment and neuromuscular junction formation.
Molecular metabolism (2017-11-22)
Richard Mills, Hermes Taylor-Weiner, Jorge C Correia, Leandro Z Agudelo, Ilary Allodi, Christina Kolonelou, Vicente Martinez-Redondo, Duarte M S Ferreira, Susanne Nichterwitz, Laura H Comley, Vanessa Lundin, Eva Hedlund, Jorge L Ruas, Ana I Teixeira
ABSTRACT
We examined whether skeletal muscle overexpression of PGC-1α1 or PGC-1α4 affected myokine secretion and neuromuscular junction (NMJ) formation. A microfluidic device was used to model endocrine signaling and NMJ formation between primary mouse myoblast-derived myotubes and embryonic stem cell-derived motor neurons. Differences in hydrostatic pressure allowed for fluidic isolation of either cell type or unidirectional signaling in the fluid phase. Myotubes were transduced to overexpress PGC-1α1 or PGC-1α4, and myokine secretion was quantified using a proximity extension assay. Morphological and functional changes in NMJs were measured by fluorescent microscopy and by monitoring muscle contraction upon motor neuron stimulation. Skeletal muscle transduction with PGC-1α1, but not PGC-1α4, increased NMJ formation and size. PGC-1α1 increased muscle secretion of neurturin, which was sufficient and necessary for the effects of muscle PGC-1α1 on NMJ formation. Our findings indicate that neurturin is a mediator of PGC-1α1-dependent retrograde signaling from muscle to motor neurons.
Sigma-Aldrich
(+)-Tubocurarine chloride pentahydrate, ≥97.0% (TLC)
Sigma-Aldrich
Monoclonal Anti-Green Fluorescent Protein (GFP) antibody produced in mouse, clone GFP-20, ascites fluid
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 10,000