Merck
  • Reaction products of [60]fullerene during the autoxidation of methyl linoleate in bulk phase.
Reaction products of [60]fullerene during the autoxidation of methyl linoleate in bulk phase.
Chemistry and physics of lipids (2012-01-03)
Akio Kadowaki, Satoshi Iwamoto, Ryo Yamauchi
ABSTRACT
The antioxidative action of fullerenes has received much attention, but their reaction mechanism toward lipid-derived peroxyl radicals has not been well elucidated. In this study, the reaction products of [60]fullerene (C(60)) during the autoxidation of methyl linoleate (MeL) were isolated and their structures were characterized. MeL containing 0.1mol% C(60) was autoxidized at 60°C in bulk phase and two reaction products of C(60), 1 and 2, were obtained. The structure of 1 was the addition products of C(60) with 9-peroxyl-10-alkyl radicals of methyl (11E)-13-hydroperoxy-11-octadecaenoate (1a and 1b) and with 12-alkyl-13-peroxyl radicals of methyl (10E)-9-hydroperoxy-10-octadecaenoate (1c and 1d). 2 was a mixture of the addition products of C(60) with 9,11-dialkyl radicals of methyl 9,12-octadecadienoate (2a) and with 11,13-dialkyl radicals of methyl 9,12-octadecadienoate (2b). When MeL containing 0.1mol% C(60) was autoxidized at 60°C under air-sufficient and air-insufficient conditions, C(60) could suppress the formation of MeL hydroperoxides in both conditions. The reaction product of C(60) first formed was 2 even under air-sufficient conditions, and then 1 was accumulated. The results indicate that the primary antioxidative action of C(60) would be trapping of chain-initiating carbon-centered radicals of unsaturated lipid to form 2.
Product Description
Supelco
cis-9,cis-12-Octadecadienoic acid methyl ester, certified reference material, 10 mg/mL in heptane
Supelco
trans-9,12-Octadecadienoic acid methyl ester, certified reference material, 10 mg/mL in heptane
Sigma-Aldrich
Methyl linolelaidate
Sigma-Aldrich
Methyl linoleate, ≥98% (GC)
Supelco
Methyl linolelaidate, analytical standard
Supelco
Methyl linoleate, analytical standard