Merck
  • The complete cell lineage and MAPK- and Otx-dependent specification of the dopaminergic cells in the Ciona brain.
The complete cell lineage and MAPK- and Otx-dependent specification of the dopaminergic cells in the Ciona brain.
Development (Cambridge, England) (2021-06-15)
Kouhei Oonuma, Takehiro G Kusakabe
ABSTRACT
The Ciona larva has served as a unique model for understanding the development of dopaminergic cells at single-cell resolution owing to the exceptionally small number of neurons in its brain and its fixed cell lineage during embryogenesis. A recent study suggested that the transcription factors Fer2 and Meis directly regulate the dopamine synthesis genes in Ciona, but the dopaminergic cell lineage and the gene regulatory networks that control the development of dopaminergic cells have not been fully elucidated. Here, we reveal that the dopaminergic cells in Ciona are derived from a bilateral pair of cells called a9.37 cells at the center of the neural plate. The a9.37 cells divide along the anterior-posterior axis, and all of the descendants of the posterior daughter cells differentiate into the dopaminergic cells. We show that the MAPK pathway and the transcription factor Otx are required for the expression of Fer2 in the dopaminergic cell lineage. Our findings establish the cellular and molecular framework for fully understanding the commitment to dopaminergic cells in the simple chordate brain.
Sigma-Aldrich
SU-5402, ≥98% (HPLC)
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder