Merck
  • The cellular prion protein interacts with and promotes the activity of Na,K-ATPases.
The cellular prion protein interacts with and promotes the activity of Na,K-ATPases.
PloS one (2021-12-01)
Declan Williams, Mohadeseh Mehrabian, Hamza Arshad, Shehab Eid, Christopher Sackmann, Wenda Zhao, Xinzhu Wang, Farinaz Ghodrati, Claire E Verkuyl, Joel C Watts, Gerold Schmitt-Ulms
ABSTRACT
The prion protein (PrP) is best known for its ability to cause fatal neurodegenerative diseases in humans and animals. Here, we revisited its molecular environment in the brain using a well-developed affinity-capture mass spectrometry workflow that offers robust relative quantitation. The analysis confirmed many previously reported interactions. It also pointed toward a profound enrichment of Na,K-ATPases (NKAs) in proximity to cellular PrP (PrPC). Follow-on work validated the interaction, demonstrated partial co-localization of the ATP1A1 and PrPC, and revealed that cells exposed to cardiac glycoside (CG) inhibitors of NKAs exhibit correlated changes to the steady-state levels of both proteins. Moreover, the presence of PrPC was observed to promote the ion uptake activity of NKAs in a human co-culture paradigm of differentiated neurons and glia cells, and in mouse neuroblastoma cells. Consistent with this finding, changes in the expression of 5'-nucleotidase that manifest in wild-type cells in response to CG exposure can also be observed in untreated PrPC-deficient cells. Finally, the endoproteolytic cleavage of the glial fibrillary acidic protein, a hallmark of late-stage prion disease, can also be induced by CGs, raising the prospect that a loss of NKA activity may contribute to the pathobiology of prion diseases.
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Calpain Inhibitor I, ≥97% (TLC), powder
Sigma-Aldrich
Anti-Sodium Pump Subunit alpha-2 Antibody, from rabbit, purified by affinity chromatography
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial