Merck
  • Longitudinal Pathogenic Properties and N-Glycosylation Profile of Antibodies from Patients with Pemphigus after Corticosteroid Treatment.
Longitudinal Pathogenic Properties and N-Glycosylation Profile of Antibodies from Patients with Pemphigus after Corticosteroid Treatment.
Biomedicines (2021-10-24)
Marie Petit, Marie-Laure Walet-Balieu, Damien Schapman, Marie-Laure Golinski, Carole Burel, Marion Barray, Laurent Drouot, Maud Maho-Vaillant, Vivien Hébert, Olivier Boyer, Muriel Bardor, Pascal Joly, Sébastien Calbo
ABSTRACT
Pemphigus vulgaris is an autoimmune disease that occurs due to pathogenic autoantibodies that recognize the following epidermal adhesion proteins: desmogleins. Systemic corticosteroids usually decrease the titers of anti-desmoglein autoantibodies and improve patients' conditions. Since modifications of IgG N-glycosylation have been described in some autoimmune diseases, we hypothesized that changes in the pathogenic activity of pemphigus IgG could be related to changes in their N-glycosylation profile. The purpose of this study was to assess, longitudinally, the pathogenicity of pemphigus serum IgG and their N-glycosylation profile during phases of disease activity and clinical remission. The pathogenic activity of serum IgG was measured in vitro on immortalized keratinocytes, by immunofluorescence and dissociation assays, and IgG N-glycans were analyzed by mass spectrometry. We showed (i) a correlation between pemphigus clinical activity and the pathogenicity of serum IgG at baseline and at month 6, while the persistence of the in vitro pathogenic activity of IgG during its evolution, even in patients in clinical remission, seemed to be predictive of relapse; (ii) that modifications of the N-glycan structure were altered the in vitro pathogenicity of patients' autoantibodies; (iii) that the pathogenic properties of pemphigus IgG did not appear to be related to the disparity in IgG N-glycans during the course of pemphigus.