Merck
  • Chemerin Affects P4 and E2 Synthesis in the Porcine Endometrium during Early Pregnancy.
Chemerin Affects P4 and E2 Synthesis in the Porcine Endometrium during Early Pregnancy.
International journal of molecular sciences (2022-01-22)
Marlena Gudelska, Kamil Dobrzyn, Marta Kiezun, Katarzyna Kisielewska, Edyta Rytelewska, Tadeusz Kaminski, Nina Smolinska
ABSTRACT
Chemerin, belonging to the adipokine family, exhibits pleiotropic activity. We hypothesised that the adipokine could be involved in the regulation of steroidogenesis in the porcine endometrium. Thus, the aim of this study was to determine the effect of chemerin on the key steroidogenic enzyme proteins' abundance (Western blot), as well as on P4 and E2 secretion (radioimmunoassay) by the porcine endometrium during early pregnancy and the mid-luteal phase of the oestrous cycle. Moreover, we investigated the hormone impact on Erk and Akt signalling pathway activation (Western blot). Chemerin stimulated E2 production on days 10 to 11 of pregnancy. On days 10 to 11 and 15 to 16 of gestation, and on days 10 to 11 of the cycle, chemerin enhanced the expression of StAR and all steroidogenic enzyme proteins. On days 12 to 13 of pregnancy, chemerin decreased StAR and most of the steroidogenic enzyme proteins' abundance, whereas the P450C17 abundance was increased. On days 27 to 28 of pregnancy, chemerin increased StAR and P450C17 protein contents and decreased 3βHSD protein amounts. It was noted that the adipokine inhibited Erk1/2 and stimulated Akt phosphorylation. The obtained results indicate that chemerin affected P4 and E2 synthesis through the Erk1/2 and Akt signalling pathways.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Fc, HRP conjugate, 0.8 mg/mL, Chemicon®