- Overexpression of EphB4 promotes neurogenesis, but inhibits neuroinflammation in mice with acute ischemic stroke.
Overexpression of EphB4 promotes neurogenesis, but inhibits neuroinflammation in mice with acute ischemic stroke.
Ischemic stroke is one of the most common diseases that has a high rate of mortality, and has become a burden to the healthcare system. Previous research has shown that EPH receptor B4 (EphB4) promotes neural stem cell proliferation and differentiation in vitro. However, little is known regarding its role in the neurogenesis of ischemic stroke in vivo. Thus, the present study aimed to verify whether EphB4 was a key regulator of neurogenesis in ischemic stroke in vivo. Cerebral ischemia was induced in C57BL/6J mice via middle cerebral artery occlusion (MCAO), followed by reperfusion. Immunofluorescence staining was performed to evaluate the effect of EphB4 on the neurogenesis in cerebral cortex. The levels of inflammatory cytokines were determined using an ELISA kit. The expression levels of ABL proto‑oncogene 1, non‑receptor tyrosine kinase (ABL1)/Cyclin D1 signaling pathway‑related proteins were detected via western blotting. The current findings indicated that EphB4 expression was significantly increased in the cerebral cortex of MCAO model mice in comparison with sham‑operated mice. Moreover, EphB4 appeared to be expressed in neural stem cells (Nestin+), and persisted as these cells became neuronal progenitors (Sox2+), neuroblasts [doublecortin (DCX)+], and eventually mature neurons [neuronal nuclei (NeuN)+]. Overexpression of EphB4 elevated the number of proliferating (bromodeoxyuridine+, Ki67+) and differentiated cells (Nestin+, Sox2+, DCX+ and NeuN+), indicating the promoting effect of EphB4 on the neurogenesis of ischemic stroke. Furthermore, EphB4 overexpression alleviated the inflammation injury in MCAO model mice. The expression levels of proteins‑related to the ABL1/Cyclin D1 signaling pathway were significantly increased by the overexpression of EphB4, which suggested that restoration of EphB4 promoted the activation of the ABL1/Cyclin D1 signaling pathway. In conclusion, this study contributes to the current understanding of the mechanisms of EphB4 in exerting neurorestorative effects and may recommend a potential new strategy for ischemic stroke treatment.