Merck
  • Elevated Baseline Serum PD-L1 Level May Predict Poor Outcomes from Breast Cancer in African-American and Hispanic Women.
Elevated Baseline Serum PD-L1 Level May Predict Poor Outcomes from Breast Cancer in African-American and Hispanic Women.
Journal of clinical medicine (2022-01-22)
Yanyuan Wu, Pranabananda Dutta, Sheilah Clayton, Amaya McCloud, Jaydutt V Vadgama
ABSTRACT
The therapeutic targeting of PD-1/PD-L1 has shown clinical efficacy in treating metastatic breast cancer. We investigated the clinical significance of measuring serum PD-L1 levels in African-American and Hispanic women with breast cancer. PD-L1 levels were measured with the ELISA method from the serum samples of 244 African-Americans and Hispanics with breast cancer and 155 women without cancers. The levels of INFα2 and TNFα were measured with a Luminex multiplex assay. The protein levels of pAkt and CD44/CD24 in tumor cells were tested with immunohistochemistry analysis. Cox regression was used to assess the predicting role of serum PD-L1 for disease-free survival (DFS). PD-L1 levels were significantly elevated in breast cancer cases compared to non-cancer cases. The high PD-L1 levels were associated with HER2-positive and triple-negative breast cancer. PD-L1 level independently predicted DFS in both African-American and Hispanic women. The evaluated PD-L1 level was found to be associated with high IFNα2 and TNFα in breast cancer patients. PD-L1 serum levels can predict DFS in African American and Hispanic women with breast cancer. Furthermore, a high level of PD-L1 is more likely to be associated with tumor loss PTEN and the activation of Akt or with breast cancer cells expressing CD44high/CD24low. Further validation studies are needed to determine if PD-L1 could serve as a biomarker for patient selection for anti-PD-L1 therapy and assess treatment outcomes.