Merck
  • Exposure to High-Altitude Environment Is Associated with Drug Transporters Change: microRNA-873-5p-Mediated Alteration of Function and Expression Levels of Drug Transporters under Hypoxia.
Exposure to High-Altitude Environment Is Associated with Drug Transporters Change: microRNA-873-5p-Mediated Alteration of Function and Expression Levels of Drug Transporters under Hypoxia.
Drug metabolism and disposition: the biological fate of chemicals (2021-12-01)
Yabin Duan, Xue Bai, Jianxin Yang, Yang Zhou, Wenqi Gu, Guiqin Liu, Qian Wang, Junbo Zhu, Linli La, Xiangyang Li
ABSTRACT
Hypoxia is the main characteristic of a high-altitude environment, affecting drug metabolism. However, so far, the mechanism of microRNA (miRNA) involved in the regulation of drug metabolism and transporters under high-altitude hypoxia is still unclear. This study aims to investigate the functions and expression levels of multidrug resistance protein 1 (MDR1), multidrug resistance-associated protein 2 (MRP2), breast cancer resistance protein (BCRP), peptide transport 1 (PEPT1), and organic anion-transporting polypeptides 2B1 (OATP2B1) in rats and colon cancer (Caco-2) cells after exposure to high-altitude hypoxia. The protein and mRNA expression of MDR1, MRP2, BCRP, PEPT1, and OATP2B1 were determined by Western blot and qPCR. The functions of MDR1, MRP2, BCRP, PEPT1, and OATP2B1 were evaluated by determining the effective intestinal permeability and absorption rate constants of their specific substrates in rats under high-altitude hypoxia, and uptake and transport studies were performed on Caco-2 cells. To screen the miRNA associated with hypoxia, Caco-2 cells were examined by high throughput sequencing. We observed that the miR-873-5p was significantly decreased under hypoxia and might target MDR1 and pregnane X receptor (PXR). To clarify whether miR-873-5p regulates MDR1 and PXR under hypoxia, Caco-2 cells were transfected with mimics or inhibitors of miR-873-5p and negative control (NC). The function and expression of drug transporters were found to be significantly increased in rats and Caco-2 cells under hypoxia. We found that miR-873-5p regulated MDR1 and PXR expression. Herein, it is shown that miRNA may affect the expression of drug transporter and nuclear receptor under hypoxia. SIGNIFICANCE STATEMENT: This study explores if alterations to the microRNAs (miRNAs), induced by high-altitude hypoxia, can be translated to altered drug transporters. Among miRNAs, which show a significant change in a hypoxic environment, miR-873-5p can act on the multidrug resistance protein 1 (MDR1) gene; however, there are multiple miRNAs that can act on the pregnane X receptor (PXR). This study speculates that the miRNA-PXR-drug transporter axis is important in the physiological disposition of drugs.