Merck
  • Ionic Mechanisms of Impulse Propagation Failure in the FHF2-Deficient Heart.
Ionic Mechanisms of Impulse Propagation Failure in the FHF2-Deficient Heart.
Circulation research (2020-09-24)
David S Park, Akshay Shekhar, John Santucci, Gabriel Redel-Traub, Sergio Solinas, Shana Mintz, Xianming Lin, Ernest Whanwook Chang, Deven Narke, Yuhe Xia, Mitchell Goldfarb, Glenn I Fishman
ABSTRACT
FHFs (fibroblast growth factor homologous factors) are key regulators of sodium channel (NaV) inactivation. Mutations in these critical proteins have been implicated in human diseases including Brugada syndrome, idiopathic ventricular arrhythmias, and epileptic encephalopathy. The underlying ionic mechanisms by which reduced Nav availability in Fhf2 knockout (Fhf2KO) mice predisposes to abnormal excitability at the tissue level are not well defined. Using animal models and theoretical multicellular linear strands, we examined how FHF2 orchestrates the interdependency of sodium, calcium, and gap junctional conductances to safeguard cardiac conduction. Fhf2KO mice were challenged by reducing calcium conductance (gCaV) using verapamil or by reducing gap junctional conductance (Gj) using carbenoxolone or by backcrossing into a cardiomyocyte-specific Cx43 (connexin 43) heterozygous background. All conditions produced conduction block in Fhf2KO mice, with Fhf2 wild-type (Fhf2WT) mice showing normal impulse propagation. To explore the ionic mechanisms of block in Fhf2KO hearts, multicellular linear strand models incorporating FHF2-deficient Nav inactivation properties were constructed and faithfully recapitulated conduction abnormalities seen in mutant hearts. The mechanisms of conduction block in mutant strands with reduced gCaV or diminished Gj are very different. Enhanced Nav inactivation due to FHF2 deficiency shifts dependence onto calcium current (ICa) to sustain electrotonic driving force, axial current flow, and action potential (AP) generation from cell-to-cell. In the setting of diminished Gj, slower charging time from upstream cells conspires with accelerated Nav inactivation in mutant strands to prevent sufficient downstream cell charging for AP propagation. FHF2-dependent effects on Nav inactivation ensure adequate sodium current (INa) reserve to safeguard against numerous threats to reliable cardiac impulse propagation.
Sigma-Aldrich
Donkey serum
Roche
Liberase TM Research Grade, medium Thermolysin concentration
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder