Merck
  • Phiclust: a clusterability measure for single-cell transcriptomics reveals phenotypic subpopulations.
Phiclust: a clusterability measure for single-cell transcriptomics reveals phenotypic subpopulations.
Genome biology (2022-01-12)
Maria Mircea, Mazène Hochane, Xueying Fan, Susana M Chuva de Sousa Lopes, Diego Garlaschelli, Stefan Semrau
ABSTRACT
The ability to discover new cell phenotypes by unsupervised clustering of single-cell transcriptomes has revolutionized biology. Currently, there is no principled way to decide whether a cluster of cells contains meaningful subpopulations that should be further resolved. Here, we present phiclust (ϕclust), a clusterability measure derived from random matrix theory that can be used to identify cell clusters with non-random substructure, testably leading to the discovery of previously overlooked phenotypes.