Merck
  • Structure-activity relationship studies of dynorphin A and related peptides.
Structure-activity relationship studies of dynorphin A and related peptides.
Peptides (1998-10-24)
T Naqvi, W Haq, K B Mathur
ABSTRACT
An up-to-date review is presented covering all the available information concerning the isolation, discovery, synthesis, conformation, receptor binding characteristics, pharmacological properties and SAR studies of dynorphin A and related peptides. The potential of dynorphin A and its analogs has yet to be fully realized.
Sigma-Aldrich
Dynorphin A porcine, ≥95% (HPLC)