Merck
  • Loss of Transforming Growth Factor Beta Signaling in Aortic Smooth Muscle Cells Causes Endothelial Dysfunction and Aortic Hypercontractility.
Loss of Transforming Growth Factor Beta Signaling in Aortic Smooth Muscle Cells Causes Endothelial Dysfunction and Aortic Hypercontractility.
Arteriosclerosis, thrombosis, and vascular biology (2021-04-16)
Jay Zhu, Stoyan Angelov, Ilkay Alp Yildirim, Hao Wei, Jie Hong Hu, Mark W Majesky, Frank V Brozovich, Francis Kim, David A Dichek
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Sodium diethyldithiocarbamate trihydrate
Pricing and availability is not currently available.
Sigma-Aldrich
Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
(R)-(−)-Phenylephrine hydrochloride, powder
Pricing and availability is not currently available.