Merck
  • PDIP38 is a novel mitotic spindle-associated protein that affects spindle organization and chromosome segregation.
PDIP38 is a novel mitotic spindle-associated protein that affects spindle organization and chromosome segregation.
Cell cycle (Georgetown, Tex.) (2008-10-10)
Esther Klaile, Alexander Kukalev, Björn Obrink, Mario M Müller
ABSTRACT
In order to maintain genomic integrity during mitosis, cells assemble the mitotic spindle to separate sister chromosomes to the two daughter cells. A variety of motor- and non motor-proteins are involved in the organization and regulation of this complex apparatus. DNA polymerase delta-interacting protein 38 (PDIP38) is a highly conserved protein and has so far been shown to be a cytoplasmic and nuclear protein. Cell cycle dependent nuclear localization and the interaction with DNA polymerase delta and proliferating cell nuclear antigen (PCNA) indicate a role for PDIP38 in DNA modification and/or proliferation. Here, we show for the first time that PDIP38 localizes to the mitotic spindle throughout mitosis. Using anti-PDIP38 antibody injections and siRNA silencing, we demonstrate that PDIP38 loss-of-function causes problems with spindle organization, aberrant chromosome segregation, and multinucleated cells. Taken together, the data indicate different roles for PDIP38 in safeguarding a proper cell division at various stages of the cell cycle, including DNA synthesis and repair, organization of the mitotic spindle and chromosome segregation.