Merck
  • Carcinomas assemble a filamentous CXCL12-keratin-19 coating that suppresses T cell-mediated immune attack.
Carcinomas assemble a filamentous CXCL12-keratin-19 coating that suppresses T cell-mediated immune attack.
Proceedings of the National Academy of Sciences of the United States of America (2022-01-21)
Zhikai Wang, Philip Moresco, Ran Yan, Jiayun Li, Ya Gao, Daniele Biasci, Min Yao, Jordan Pearson, Jaclyn F Hechtman, Tobias Janowitz, Raza M Zaidi, Matthew J Weiss, Douglas T Fearon
ABSTRACT
Cancer immunotherapy frequently fails because most carcinomas have few T cells, suggesting that cancers can suppress T cell infiltration. Here, we show that cancer cells of human pancreatic ductal adenocarcinoma (PDA), colorectal cancer, and breast cancer are coated with transglutaminase-2 (TGM2)-dependent covalent CXCL12-keratin-19 (KRT19) heterodimers that are organized as filamentous networks. Since a dimeric form of CXCL12 suppresses the motility of human T cells, we determined whether this polymeric CXCL12-KRT19 coating mediated T cell exclusion. Mouse tumors containing control PDA cells exhibited the CXCL12-KRT19 coating, excluded T cells, and did not respond to treatment with anti-PD-1 antibody. Tumors containing PDA cells not expressing either KRT19 or TGM2 lacked the CXCL12-KRT19 coating, were infiltrated with activated CD8+ T cells, and growth was suppressed with anti-PD-1 antibody treatment. Thus, carcinomas assemble a CXCL12-KRT19 coating to evade cancer immune attack.
Product Description
Roche
DNase I, grade II, from bovine pancreas
Sigma-Aldrich
Human Plasma Fibronectin Purified Protein, from human plasma, liquid, 1 mg/mL (100 MG pack size is lyophilized), purified protein, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture