Merck
IGF2: epigenetic regulation and role in development and disease.
Cytokine & growth factor reviews (2008-03-01)
Wendy Chao, Patricia A D'Amore
ABSTRACT
Insulin-like growth factor II (IGF2) is perhaps the most intricately regulated of all growth factors characterized to date. Its gene is imprinted--only one allele is active, depending on parental origin--and this pattern of expression is maintained epigenetically in almost all tissues. IGF2 activity is further controlled through differential expression of receptors and IGF-binding proteins (IGFBPs) that determine protein availability. This complex and multifaceted regulation emphasizes the importance of accurate IGF2 expression and activity. This review will examine the regulation of the IGF2 gene and what it has revealed about the phenomenon of imprinting, which is frequently disrupted in cancer. IGF2 protein function will be discussed, along with diseases that involve IGF2 overexpression. Roles for IGF2 in sonic hedgehog (Shh) signaling and angiogenesis will also be explored.