Merck
Bioaccessibility of folate in faba bean, oat, rye and wheat matrices.
Food chemistry (2021-02-24)
Fengyuan Liu, Susanna Kariluoto, Minnamari Edelmann, Vieno Piironen
ABSTRACT
Cereals and legumes are rich in folate. However, due to the instability of folate, processing and digestion can induce significant folate loss. In this paper, folate bioaccessibility of faba bean, oat, rye and wheat flours and pastes was studied using a static in vitro digestion model. Folate bioaccessibility depended on food matrices, varying from 42% to 67% in flours and from 40% to 123% in pastes. Digestion was associated with the interconversion of formyl folates, as well as the increase of oxidised vitamers and decrease of reduced vitamers. Especially in faba bean, 5-methyltetrahydrofolate showed surprisingly good stability both in digestion and heat treatment, resulting in high bioaccessibility. The physiological concentration of ascorbic acid did not stabilise folate in digestion; however, a higher level helped to maintain reduced vitamers. Heat treatment (10-min paste making) could improve folate bioaccessibility by liberating folate from the food matrices and by altering folate vitamer distribution.
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
Sigma-Aldrich
Trypsin from porcine pancreas, Type IX-S, lyophilized powder, 13,000-20,000 BAEE units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Protease from Streptomyces griseus, powder, BioReagent, suitable for mouse embryo cell culture, ≥3.5 units/mg solid