- Anaerobic production of alginate by Pseudomonas aeruginosa: alginate restricts diffusion of oxygen.
Anaerobic production of alginate by Pseudomonas aeruginosa: alginate restricts diffusion of oxygen.
Journal of bacteriology (1996-12-01)
D J Hassett
PMID8955420
ABSTRACT
Pseudomonas aeruginosa produced alginate and elevated algD (encoding GDPmannose 6-dehydrogenase) transcription under strict anaerobic conditions, especially when using nitrate as a terminal electron acceptor. Purified alginate added to bacterial suspensions caused a decrease in growth, suggesting that alginate contributes to oxygen limitation for the organism and likely for patients afflicted with the inherited autosomal disease cystic fibrosis.