Merck
  • An autophagy-related protein Becn2 regulates cocaine reward behaviors in the dopaminergic system.
An autophagy-related protein Becn2 regulates cocaine reward behaviors in the dopaminergic system.
Science advances (2021-02-21)
Yoon-Jin Kim, Qingyao Kong, Soh Yamamoto, Kenta Kuramoto, Mei Huang, Nan Wang, Jung Hwa Hong, Tong Xiao, Beth Levine, Xianxiu Qiu, Yanxiang Zhao, Richard J Miller, Hongxin Dong, Herbert Y Meltzer, Ming Xu, Congcong He
ABSTRACT
Drug abuse is a foremost public health problem. Cocaine is a widely abused drug worldwide that produces various reward-related behaviors. The mechanisms that underlie cocaine-induced disorders are unresolved, and effective treatments are lacking. Here, we found that an autophagy-related protein Becn2 is a previously unidentified regulator of cocaine reward behaviors. Becn2 deletion protects mice from cocaine-stimulated locomotion and reward behaviors, as well as cocaine-induced dopamine accumulation and signaling, by increasing presynaptic dopamine receptor 2 (D2R) autoreceptors in dopamine neurons. Becn2 regulates D2R endolysosomal trafficking, degradation, and cocaine-induced behaviors via interacting with a D2R-bound adaptor GASP1. Inactivating Becn2 by upstream autophagy inhibitors stabilizes striatal presynaptic D2R, reduces dopamine release and signaling, and prevents cocaine reward in normal mice. Thus, the autophagy protein Becn2 is essential for cocaine psychomotor stimulation and reward through regulating dopamine neurotransmission, and targeting Becn2 by autophagy inhibitors is a potential strategy to prevent cocaine-induced behaviors.
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Beclin-2, from rabbit
Sigma-Aldrich
Anti-Dopamine Transporter Antibody, NT, clone DAT-Nt, culture supernatant, clone DAT-Nt, Chemicon®
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit