Merck
  • Nephroprotective Role of Chrysophanol in Hypoxia/Reoxygenation-Induced Renal Cell Damage via Apoptosis, ER Stress, and Ferroptosis.
Nephroprotective Role of Chrysophanol in Hypoxia/Reoxygenation-Induced Renal Cell Damage via Apoptosis, ER Stress, and Ferroptosis.
Biomedicines (2021-09-29)
Chih-Hung Lin, Han-Fang Tseng, Po-Chun Hsieh, Valeria Chiu, Ting-Yun Lin, Chou-Chin Lan, I-Shiang Tzeng, Huan-Nung Chao, Chia-Chen Hsu, Chan-Yen Kuo
ABSTRACT
Acute kidney injury (AKI) is caused by hypoxia-reoxygenation (H/R), which is a kidney injury produced by a variety of causes, resulting in the remaining portion of the kidney function being unable to maintain the balance for performing the tasks of waste excretion metabolism, and electrolyte and acid-base balance. Many studies have reported the use of Chinese medicine to slow down the progression and alleviate the complications of chronic renal failure. Chrysophanol is a component of Rheum officinale Baill, a traditional Chinese medicine that has been clinically used to treat renal disease. We aimed to study the nephroprotective effect of chrysophanol on hypoxia/ reoxygenation (H/R)-induced cell damage. The results showed that chrysophanol prevented H/R-induced apoptosis via downregulation of cleaved Caspase-3, p-JNK, and Bax but upregulation of Bcl-2 expression. In contrast, chrysophanol attenuated H/R-induced endoplasmic reticulum (ER) stress via the downregulation of CHOP and p-IRE1α expression. Our data demonstrated that chrysophanol alleviated H/R-induced lipid ROS accumulation and ferroptosis. Therefore, we propose that chrysophanol may have a protective effect against AKI by regulating apoptosis, ER stress, and ferroptosis.
Sigma-Aldrich
Phosphate buffered saline, 10× concentrate, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
Cell Freezing Medium-DMSO 1×, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)