Merck
  • Collagen-based fibrillar multilayer films cross-linked by a natural agent.
Collagen-based fibrillar multilayer films cross-linked by a natural agent.
Biomacromolecules (2012-06-06)
Christophe Chaubaroux, Engin Vrana, Christian Debry, Pierre Schaaf, Bernard Senger, Jean-Claude Voegel, Youssef Haikel, Christian Ringwald, Joseph Hemmerlé, Philippe Lavalle, Fouzia Boulmedais
ABSTRACT
Surface functionalization plays an important role in the design of biomedical implants, especially when layer forming cells, such as endothelial or epithelial cells, are needed. In this study, we define a novel nanoscale surface coating composed of collagen/alginate polyelectrolyte multilayers and cross-linked for stability with genipin. This buildup follows an exponential growth regime versus the number of deposition cycles with a distinct nanofibrillar structure that is not damaged by the cross-linking step. Stability and cell compatibility of the cross-linked coatings were studied with human umbilical vein endothelial cells. The surface coating can be covered by a monolayer of vascular endothelial cells within 5 days. Genipin cross-linking renders the surface more suitable for cell attachment and proliferation compared to glutaraldehyde (more conventional cross-linker) cross-linked surfaces, where cell clumps in dispersed areas were observed. In summary, it is possible with the defined system to build fibrillar structures with a nanoscale control of film thickness, which would be useful for in vivo applications such as inner lining of lumens for vascular and tracheal implants.
Product Description
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Resazurin based